Tetramethrin mixture of isomers 98+%

Supplier: Thermo Fisher Scientific
Warning

471880010 471880050
471880010.EA 50 EUR
471880010. 471880050.
Tetramethrin mixture of isomers 98+%
Tetramethrin
Formula: C₁₉H₂₅NO₄
Boiling Pt: 453,2°C (760mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00084635
CAS Number: 7696-12-0
EINECS: 231-711-6
UN: 3077
ADR: 9,III
Merck Index: 13,09296

Order Now

Specification Test Results

Appearance white to light yellow to light beige powder or crystals
Infrared spectrum Conforms
HPLC ≥98.0 % (mix of cis/trans)
Melting point 74°C to 81°C
Water ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR