Octaethylene glycol 97%

Supplier: Thermo Fisher Scientific

471960010 471960050
471960010.EA 90.5 EUR
471960010. 471960050.
Octaethylene glycol 97%
Octaethylene glycol
Formula: C₁₆H₃₄O₉
Storage Temperature: Ambient
MDL Number: MFCD00698694
CAS Number: 5117-19-1
EINECS: 225-856-4

Order Now

Specification Test Results

Appearance colorless to light yellow or white liquid or solid
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR