Benzyltoluene 97%

Supplier: Thermo Fisher Scientific

473430010 473432500
473430010.EA 106 EUR
473430010. 473432500.
Benzyltoluene 97%
Benzyltoluene
Formula: C₁₄H₁₄
MDL Number: MFCD00048102
CAS Number: 27776-01-8

Order Now

Specification Test Results

Appearance colorless liquid
Infrared spectrum Conforms
HPLC ≥96 % (total isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR