Hippuric acid, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Benzoyl amidoacetic acid, Benzoylaminoacetic acid, Bz-Gly-OH, Benzoyl glycocoll, 2-Benzamidoacetic acid, N-Benzoylglycine

M-Clarity™ quality level = MQ200

8.20649.0100 8.20649.0500
8.20649.0100EA 58.2 EUR
8.20649.0100 8.20649.0500
Hippuric acid, Sigma-Aldrich®
Hippuric acid
Formula: C₆H₅C(O)NHCH₂COOH
MW: 179.18 g/mol
Boiling Pt: 399
Melting Pt: 271
Density: 1.308 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002692
CAS Number: 495-69-2
EINECS: 207-806-3
Merck Index: 13,04735

Order Now

Specification Test Results

Solubility 3.26 g/l (20 °C)
Melting Point 188 - 190 °C
Molar Mass 179.17 g/mol
Bulk Density 630 kg/m3
pH 2.5 - 3.5 (10 g/l, H2O, 20 °C) (slurry)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR