1,1,3,3-Tetramethylbutylamine 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2,4,4-Trimethylpentan-2-amine, 2-Amino-2,4,4-trimethylpentane, tert-Octylamine
129500050
129505000
ACRO129500050EA
25.3
EUR
ACRO129500050
ACRO129505000
1,1,3,3-Tetramethylbutylamine 95%
1,1,3,3-Tetramethylbutylamine
Formula:
C₈H₁₉N Boiling Pt: 137 °C (1013 hPa) Melting Pt: –67 °C Density: 0,805 g/cm³ Flash Pt: 32 °C |
MDL Number:
MFCD00008053 CAS Number: 107-45-9 EINECS: 203-491-1 UN: 2734 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4235 to 1.4255 (20°C, 589 nm) |
Water | ≤0.3 % |
GC | ≥94 % |
Color scale | ≤25 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...