Conical Funnel with Strainers
Supplier: Adelphi Pharma Hygiene Products
FUN4010P FUN4012P FUN4025P
COLDFUN4010PEA 76.8 EUR
COLDFUN4010P COLDFUN4012P COLDFUN4025P
Conical Funnel with Strainers
Funnels Powder Funnels
A range of stainless steel funnels with built-in strainers which meet international pharma and food quality compliance regulations.
  • 304 grade stainless steel
  • Hygienic one-piece body construction
  • Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
  • Optional material grade and surface finish certification available upon request
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information.