1-(4-Nitrophenyl)glycerol ≥99%
Supplier: Thermo Scientific
Synonyms:
PNPG, 1-(4-Nitrophenyl)-1,2,3-propanetriol
L19968.03
L19968.06
L19968.MD
L19968.03EA
213
EUR
L19968.03
L19968.06
L19968.MD
1-(4-Nitrophenyl)glycerol ≥99%
1-(4-Nitrophenyl)glycerol
Purity: 99%
CAS No.: 2207-68-3
Molecular Formula: C9H11NO5
Formula Weight: 213.19
Odour: Odourless
Melting Point: 95-97°
Beilstein Reference: 7376091
MDL No.: MFCD00017037
Formula:
C₉H₁₁NO₅ MW: 213,19 g/mol Melting Pt: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017037 CAS Number: 2207-68-3 EINECS: 218-624-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...