Sodium tetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Sodium fluoroborate
M-Clarity™ quality level = MQ200
8.43877.0250
8.43877.1000
8.43877.9999
8.43877.0250EA
64.8
EUR
8.43877.0250
8.43877.1000
8.43877.9999
Sodium tetrafluoroborate, Sigma-Aldrich®
Sodium tetrafluoroborate
Formula:
BF₄Na MW: 109,79 g/mol Melting Pt: 384 °C Density: ∼2,47 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003515 CAS Number: 13755-29-8 EINECS: 237-340-6 UN: 1759 ADR: 8,II Merck Index: 12,08761 |
Specification Test Results
Melting Point | 384 °C |
Molar Mass | 109.79 g/mol |
Hill Formula | BF4Na |
Chemical Formula | NaBF4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...