Alizarine ≥94%

Supplier: Thermo Scientific
Warning

Synonyms: Alizarin, 1,2-Dihydroxy-9,10-anthraquinone, 1,2-Dihydroxy-9,10-anthracenedione, 1,2-Dihydroxyanthraquinone

A14404.36 A14404.0E A14404.22
A14404.36EA 247 EUR
A14404.36 A14404.0E A14404.22
Alizarine ≥94%
Alizarine

Purity: 94% CAS No.: 72-48-0 Molecular Formula: C14H8O4 Formula Weight: 240.21 Melting Point: 286-290° Boiling Point: ca 430° subl. Merck Reference: 14,251 MDL No.: MFCD00001201

Formula: C₁₄H₈O₄
MW: 240,22 g/mol
Boiling Pt: ∼430 °C (subl.)
Melting Pt: 286…290 °C
Density: 1,54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001201
CAS Number: 72-48-0
EINECS: 200-782-5
Merck Index: 14,251

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR