Xylenol orange tetrasodium salt ACS

Supplier: Thermo Scientific

41379.04 41379.09 41379.18
41379.04EA 81.4 EUR
41379.04 41379.09 41379.18
Xylenol orange tetrasodium salt ACS
Xylenol orange tetrasodium salt

CAS No.: 3618-43-7 Molecular Formula: C31H28N2Na4O13S Formula Weight: 760.59 Physical Form: Powder Odour: Odourless Melting Point: 210° dec. MDL No.: MFCD00044293

Formula: C₃₁H₂₈N₂Na₄O₁₃S
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR