3-Aminophthalic acid 95%

Supplier: Thermo Scientific
Warning

375750010 375750050
ACRO375750010EA 70.7 EUR
ACRO375750010 ACRO375750050
3-Aminophthalic acid 95%
3-Aminophthalic acid
Formula: C₈H₇NO₄
Melting Pt: 170…180 °C
CAS Number: 5434-20-8
EINECS: 226-595-9

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
GC ≥94 %
Titration with NaOH 94 to 106 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR