Carboxymethyl cellulose sodium salt
Supplier: Roth Carl
Synonyms:
Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC
6190.1
6190.2
ROTH6190.1EA
38.6
EUR
ROTH6190.1
ROTH6190.2
Carboxymethyl cellulose sodium salt
Carboxymethyl cellulose sodium salt
Formula:
[C₆H₇O₂(OH)x(OCH₂COONa)y]n Storage Temperature: Ambient |
MDL Number:
MFCD00081472 CAS Number: 9004-32-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...