1-Ethylquinaldinium iodide ≥97%
Supplier: Thermo Fisher Scientific
Synonyms:
1-Ethyl-2-methylquinolin-1-ium iodide, 1-Ethyl-2-methylquinolinium iodide
A10584.22
A10584.14
A10584.06
A10584.22EA
547
EUR
A10584.22
A10584.14
A10584.06
1-Ethylquinaldinium iodide ≥97%
1-Ethylquinaldinium iodide
Purity: 97%
CAS No.: 606-55-3
Molecular Formula: C12H14IN
Formula Weight: 299.16
Melting Point: 231° dec.
MDL No.: MFCD00031829
Formula:
C₁₂H₁₄IN Melting Pt: 231 °C (dec.) Density: 1,689 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00031829 CAS Number: 606-55-3 EINECS: 210-120-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...