Triacetoxymethylsilane 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Triacetoxy(methyl)silane, Methylsilanetriyl triacetate
B23974.09
B23974.18
B23974.30
B23974.09EA
25.7
EUR
B23974.09
B23974.18
B23974.30
Triacetoxymethylsilane 96%
Triacetoxymethylsilane
Purity: 96%
CAS No.: 4253-34-3
Molecular Formula: C7H12O6Si
Formula Weight: 220.25
Flash Point: 85°(185°F)
Melting Point: 40-45°
Boiling Point: 87-88°/3mm
Beilstein Reference: 1788668
UN No.: 1759
MDL No.: MFCD00008694
Formula:
C₇H₁₂O₆Si Boiling Pt: 87…88 °C (3 mmHg) Melting Pt: 40…45 °C Density: 1,175 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008694 CAS Number: 4253-34-3 EINECS: 224-221-9 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...