6-Bromoquinaldine 97

Supplier: Thermo Fisher Scientific
Warning

H27447.14 H27447.06
H27447.14EA 216 EUR
H27447.14 H27447.06
6-Bromoquinaldine 97
6-Bromoquinaldine

Purity: 97% CAS No.: 877-42-9 Molecular Formula: C10H8BrN Formula Weight: 222.08 Melting Point: 100-103° MDL No.: MFCD00079724

Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00079724
CAS Number: 877-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR