3-Nitro-1H-1,2,4-triazole 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Nitro-1,2,4-1H-triazole, 3-Nitro-1,2,4-triazole
L06845.03
L06845.06
L06845.03EA
49.8
EUR
L06845.03
L06845.06
3-Nitro-1H-1,2,4-triazole 96%
3-Nitro-1H-1,2,4-triazole
Purity: 96%
CAS No.: 24807-55-4
Molecular Formula: C2H2N4O2
Formula Weight: 114.06
Melting Point: ca 215° dec.
Beilstein Reference: 607167
Formula:
C₂H₂N₄O₂ MW: 114,06 g/mol Melting Pt: ∼ 215 °C (dec.) Density: 1,65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009749 CAS Number: 24807-55-4 EINECS: 246-468-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...