1,2,4-Triazole, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ200
8.08388.0025
8.08388.0100
8.08388.9025
8.08388.0025EA
44.5
EUR
8.08388.0025
8.08388.0100
8.08388.9025
1,2,4-Triazole, Sigma-Aldrich®
1,2,4-Triazole
Formula:
C₂H₃N₃ Boiling Pt: 260 °C (1013 hPa) Melting Pt: 116…119 °C Density: 1,394 g/cm³ (25 °C) |
MDL Number:
MFCD00005228 CAS Number: 288-88-0 EINECS: 206-022-9 Merck Index: 12,09735 |
Specification Test Results
Solubility | 547 g/l (22 °C) |
Melting Point | 119 - 122 °C |
Molar Mass | 69.07 g/mol |
Bulk Density | 550 kg/m3 |
Boiling Point | 259 - 261 °C (1013 hPa) |
Density | 1.15 g/cm3 (130 °C) |
pH | 8 (10 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...