Pouches for gas/steam sterilisation, Convertors®, CareFusion
Supplier: CareFusion
90308 92510 92515 92713
489-0085EA 0 EUR
489-0085 489-0071 489-0072 489-0073
Pouches for gas/steam sterilisation, Convertors®, CareFusion
Bags Sterilisation Pouches
Peel-open pape/film pouches are designed for quick, easy presentation of sterile items. The reverse side is made of tough, moisture-resistant, surgical-grade paper, which provides high porosity for fast sterilisation while maintaining an effective bacterial barrier. Heat-seal pouches can be heat-sealed or taped shut. Self-seal pouches feature a pre-folded, pressure-sensitive closure to ensure the integrity of the seal.
  • Can be used in steam or EO sterilisation
  • Chemical process indicators on every pouch identify the sterilisation process used
  • Items within the pouch are easily viewed through the strong blue-tinted 2 mm thick film side