Knitted gloves, nylon
Supplier: Bennett Safetywear
1500050021 1500010011
112-3901EA 161 EUR
112-3901 112-3900
Knitted gloves, nylon
Gloves
This fine, lightweight and close fitting nylon glove offers excellent dexterity and basic protection from abrasion, cut, tear and puncture hazards.
  • Washable
  • Dust and lint free
  • In two sizes, suitable for male or female
Colour: Beige
EN 288, 6.1-6.4 (2141)
Delivery Information: Supplied in pairs.
Certifications: EN 388 (2141)