Serological pipettes, Falcon®
Supplier: Corning
357543_COS 357550_COS 357525_COS
BDAA357543_COSEA 0 EUR
BDAA357543_COS BDAA357550_COS BDAA357525_COS
Serological pipettes, Falcon®
Pipettes Serological Pipettes
PS, transparent, graduated, sterile.
  • Coloured top with cotton plug and coloured packaging for easy identification
  • Negative graduations for extra capacity and reverse graduations (except 1 ml)
  • Non pyrogenic
Packaging: Individually wrapped in paper-peelable bags or bulk packed in bags.