VWR®, NMR Sample Tube Caps
Supplier: VWR Collection
634-0466EA 10.3 EUR
634-0466 634-0470 634-0472 634-0473 634-0478 634-0482
VWR®, NMR Sample Tube Caps
Closures Test/Sample Tube Closures
Manufactured in ethylene vinyl acetate (EVA).
  • Caps for 5 mm OD NMR tubes
  • Available in eight colours
Caps for 5 mm O.Ø NMR tubes.
Ordering information: Available in different colours.
Caution: Not recommended for use with chloroform and DMSO.