Tubing, silicone, Syndev®
Supplier: SF MEDICAL
SFM1-1050 SFM1-1350 SFM1-1550 SFM1-2950 SFM1-3350 SFM1-4050 SFM1-4250 SFM1-4450 SFM1-4650 SFM1-4850 SFM1-4950 SFM1-5250 SFM1-5450 SFM1-6050 SFM1-6250 SFM1-6450 SFM1-6850 SFM1-7050
228-0228EA 209 EUR
228-0228 228-0229 228-0230 228-0234 228-0237 228-0239 228-0240 228-0241 228-0242 228-0243 228-0244 228-0246 228-0247 228-0248 228-0249 228-0250 228-0251 228-0252
Tubing, silicone, Syndev®
Tubing Transfer Tubing
Silicone, translucent, food grade.
  • Peroxide cured
  • Manufactured to GMP in a clean room, FDA compliant to food grade regulation 177.2600 and certified to the biocompatibility requirements of USP Class VI
Suitable for peristaltic pumps, endoscopy, laparoscopy, most food processing applications and many other non critical medical, laboratory, pharmaceutical and biotechnical applications.
Temperature resistance: −60 to +220 °C
Hardness (Shore durometer A): 60 ±5