Bismarck brown Y

Supplier: Roth Carl

Synonyms: C.I. 21000, 1,3-Bis(2,3-diaminophenylazo)benzene dihydrochloride, Bismarck Brown Y (G), Basic Brown 1, Vesuvine, Phenylene Brown, Leather Brown, Basic Brown G, GX, GXP, Manchester Brown, 4,4'-[1,3-Phenylenebis(azo)]bisbenzene-1,3-diamine dihydrochloride

0313.1 0313.2
ROTH0313.1EA 31.7 EUR
ROTH0313.1 ROTH0313.2
Bismarck brown Y
Bismarck Brown Y
Formula: C₁₈H₁₈N8·2HCl
MW: 419,32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012977
CAS Number: 10114-58-6
EINECS: 233-314-3
Merck Index: 12,01294

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR