Indigo

Supplier: Roth Carl
Warning

Synonyms: Pigment Blue 66, Vat Blue 1, Indigotin, 2,2'-Bis(2,3-dihydro-3-oxoindolyliden), Indigo (synthetic), 2-(1,3-Dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-3H-indol-3-one

3903.1 3903.2
ROTH3903.1EA 0 EUR
ROTH3903.1 ROTH3903.2
Indigo
Indigo
Formula: C₁₆H₁₀N₂O₂
MW: 262.27 g/mol
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005722
CAS Number: 482-89-3
EINECS: 207-586-9
Merck Index: 12,04977

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR