Azomethine H sodium salt hydrate ~95%, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Warning

Synonyms: Sodium 5-hydroxy-4-((2-hydroxybenzylidene)amino)-7-sulphonaphthalene-2-sulphonate xhydrate, Azomethine-H monosodium salt hydrate, 8-Hydroxy-1-(salicylideneamino)naphthalene-3,6-disulphonic acid 6-sodium salt hydrate, Sodium hydrogen 4-hydroxy-5-(salicylideneamino)naphthalene-2,7-disulphonate hydrate, Azomethine-H sodium hydrate

M-Clarity™ quality level = MQ200

A3144-1G A3144-25G A3144-5G
A3144-1GEA 66.6 EUR
A3144-1G A3144-25G A3144-5G
Azomethine H sodium salt hydrate ~95%, Sigma-Aldrich®
Azomethine H sodium salt hydrate

Azomethine-H is used as a color development agent. It is used for the determination of boron, mainly in soil samples. Azomethine-H monosodium salt hydrate has been used for the colorimetric determination of boron in soil and plants.

Formula: C₁₇H₁₂NNaO₈S₂·xH₂O
MW: 463.42 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00149588
CAS Number: 206752-32-1
EINECS: 227-698-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=039fa8fabf130be6b193fa11648e1ec2d7bc445c&vid=3964aa9c3af9f4f9f45bfc0571c0424ef21f45c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Form) Powder
Appearance (Colour) Faint Yellow to Dark Yellow and Faint Orange to Dark Orange
Solubility (Solvent) 1N NaOH
Solubility (Conc) 49.00 - 51.00 mg/mL
Solubility (Turbidity) Clear to Hazy
Solubility (Colour) Faint Yellow to Dark Yellow and Faint Orange to Dark Orange
Purity (TLC) ≥ 95.00%
NMR (Solvent) D2O
Identity by NMR Consistent
NMR (Residual Solvent 1) Ethanol
NMR (% Solvent 1) ≤ 1.00%
NMR (Residual Solvent 2) Acetone
NMR (% Solvent 2) ≤ 0.50%
UV (Solvent) Water
UV (Lambda max 1) 235 - 239 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR