Zinc carbonate basic, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
1.08808.1000
1.08808.9029
1.08808.1000EA
2130
EUR
1.08808.1000
1.08808.9029
Zinc carbonate basic, SAFC®
Zinc carbonate basic
Formula:
C₂H₆O₁₂Zn₅ Melting Pt: > 140 °C Density: 3,5 g/cm³ (20 °C) |
MDL Number:
MFCD00011519 CAS Number: 5263-02-5 EINECS: 226-076-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...