CAPS (3-(cyclohexylamino)propanesulphonic acid)
Supplier: Roth Carl
Synonyms:
3-Cyclohexylaminopropanesulphonic acid
9168.1
9168.2
9168.3
ROTH9168.1EA
28.9
EUR
ROTH9168.1
ROTH9168.2
ROTH9168.3
CAPS (3-(cyclohexylamino)propanesulphonic acid)
CAPS (3-(Cyclohexylamino)propanesulphonic acid)
Formula:
C₆H₁₁NH(CH₂)₃SO₃H MW: 221,32 g/mol Melting Pt: 324 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003837 CAS Number: 1135-40-6 EINECS: 214-492-1 Merck Index: 13,01773 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...