Xylene cyanole FF

Warning

Synonyms: C.I. 42135, Sodium hydrogen 4-[[4-(ethylamino)-m-tolyl][4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulphonate, Sodium 4-((4-(ethylamino)-3-methylphenyl)(4-(ethylimino)-3-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-sulphobenzenesulphonate, acid Blue 147

EMR249010 EMR249025
EURLEMR249010EA 0 EUR
EURLEMR249010 EURLEMR249025
Xylene cyanole FF
Xylene Cyanole FF
Formula: C₂₅H₂₇N₂NaO₆S₂
MW: 538,62 g/mol
Melting Pt: 295 °C
Storage Temperature: Ambient
MDL Number: MFCD00040657
CAS Number: 2650-17-1
EINECS: 220-167-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR