Tritylium tetrafluoroborate 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Triphenylcarbenium tetrafluoroborate

A12949.09 A12949.18 A12949.30
A12949.09EA 84.8 EUR
A12949.09 A12949.18 A12949.30
Tritylium tetrafluoroborate 97%
Tritylium tetrafluoroborate

Purity: 98% CAS No.: 341-02-6 Molecular Formula: C19H15BF4 Formula Weight: 330.13 Storage Temperature: 2°C to 8°C Odour: Odourless Beilstein Reference: 5085649 UN No.: 3261 MDL No.: MFCD00013120

Formula: C₁₉H₁₅BF₄
MW: 330.13 g/mol
Melting Pt: ∼ 207 °C (dec.)
MDL Number: MFCD00013120
CAS Number: 341-02-6
EINECS: 206-433-3
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR