Lead ≥99.9985% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Supplier: Thermo Fisher Scientific
Danger

42861.BW 42861.CB
42861.BWEA 144 EUR
42861.BW 42861.CB
Lead ≥99.9985% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Lead

Purity: 99.9985% CAS No.: 7439-92-1 Physical Form: ≈35.6g/m Appearance: Grey Odour: Odourless MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11,3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR