Lead ≥99.999% (metals basis), granules

Supplier: Thermo Fisher Scientific
Danger

42928.18 42928.30
42928.18EA 211 EUR
42928.18 42928.30
Lead ≥99.999% (metals basis), granules
Lead

Purity: 99.999% CAS No.: 7439-92-1 Odour: Odourless MDL No.: MFCD00134050

Formula: Pb
Boiling Pt: 1740 °C (1013 hPa)
Melting Pt: 327 °C
Density: 11,3437 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR