Antimony, powder

Supplier: Roth Carl
Warning

9258.1 9258.2
ROTH9258.1EA 0 EUR
ROTH9258.1 ROTH9258.2
Antimony, powder
Formula: Sb
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR