Phosphorus, red ≥98.9% (metals basis), amorphous powder -100 mesh
Supplier: Thermo Scientific
|
Danger
|
10281.36
10281.22
10281.57
10281.36EA
336
EUR
10281.36
10281.22
10281.57
Phosphorus, red ≥98.9% (metals basis), amorphous powder -100 mesh
Phosphorus
Purity: 98.9%
CAS No.: 7723-14-0
Physical Form: Powder
Appearance: Reddish-brown
UN No.: 1338
MDL No.: MFCD00133771
Formula:
P MW: 30,97 g/mol Density: ∼2,2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133771 CAS Number: 7723-14-0 EINECS: 231-768-7 UN: 1338 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...