Phosphorus, red ≥99.999% (metals basis), lumps, Puratronic®

Supplier: Thermo Fisher Scientific
Danger

10670.03 10670.18
10670.03EA 58.4 EUR
10670.03 10670.18
Phosphorus, red ≥99.999% (metals basis), lumps, Puratronic®
Phosphorus

Purity: 99.999% CAS No.: 7723-14-0 Physical Form: Solid UN No.: 1338 MDL No.: MFCD00133771

Formula: P
Density: ∼2,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133771
CAS Number: 7723-14-0
EINECS: 231-768-7
UN: 1338
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR