Lanthanum ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
|
Danger
|
10220.FF
10220.FI
10220.FFEA
379
EUR
10220.FF
10220.FI
Lanthanum ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Lanthanum
Purity: 99.9%
CAS No.: 7439-91-0
Physical Form: ≈3.9g/25x25mm, Packaged in mylar under argon
Appearance: Silver-grey
UN No.: 3208
MDL No.: MFCD00011066
Formula:
La MW: 138.91 g/mol Boiling Pt: 3464 Melting Pt: 920 Density: ∼6.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011066 CAS Number: 7439-91-0 EINECS: 231-099-0 UN: 3208 ADR: 4.3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...