Zinc 99,9997 Metals basis, wire, Ø 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific

11361.G1 11361.G5 11361.G8
11361.G1EA 224 EUR
11361.G1 11361.G5 11361.G8
Zinc 99,9997 Metals basis, wire, Ø 1.0 mm (0.04 in)
Zinc

Purity: 99.9997% CAS No.: 7440-66-6 Physical Form: ≈5.60g/m Odour: Odourless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR