Zinc 99 Metals basis, mossy 2.5 cm (0.98 in) & down
Supplier: Thermo Fisher Scientific
14629.30
14629.A1
14629.30EA
36.3
EUR
14629.30
14629.A1
Zinc 99 Metals basis, mossy 2.5 cm (0.98 in) & down
Zinc
Purity: 99%
CAS No.: 7440-66-6
Odour: Odourless
MDL No.: MFCD00011291
Formula:
Zn MW: 65.39 g/mol Boiling Pt: 908 Melting Pt: 420 Density: 7.14 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011291 CAS Number: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...