(±)-2-Heptanol 99+%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol
120370500
120370010
120372500
ACRO120370500EA
37.9
EUR
ACRO120370500
ACRO120370010
ACRO120372500
(±)-2-Heptanol 99+%
(±)-2-Heptanol
Formula:
C₇H₁₆O Boiling Pt: 160…162 °C (760 mmHg) Density: 0,8175 g/cm³ Flash Pt: 59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004587 CAS Number: 543-49-7 EINECS: 208-844-3 UN: 1987 ADR: 3,III Merck Index: 13,04682 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4200 to 1.4220 (20°C, 589 nm) |
GC | ≥99.0 % |
Specific gravity | (20°C) 0.816 to 0.819 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...