(±)-2-Heptanol ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(±)-Methyl pentyl carbinol, (±)-2-Hydroxy heptane, Heptan-2-ol, (±)-Pentyl methyl carbinol
A18391.18
A18391.30
A18391.18EA
33.8
EUR
A18391.18
A18391.30
(±)-2-Heptanol ≥98%
(±)-2-Heptanol
Purity: 98%
CAS No.: 543-49-7
Molecular Formula: C7H16O
Formula Weight: 116.20
Flash Point: 59°(138°F)
Refractive Index: 1.4210
Boiling Point: 160-162°
Beilstein Reference: 1719088
Merck Reference: 14,4662
UN No.: 1987
MDL No.: MFCD00004587
Formula:
C₇H₁₆O Boiling Pt: 160…162 °C Density: 0,819 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004587 CAS Number: 543-49-7 EINECS: 208-844-3 UN: 1987 ADR: 3,III Merck Index: 14,4662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...