(±)-1,2-Propanediol
Supplier: Roth Carl
Synonyms:
Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane
0340.1
0340.2
0340.3
ROTH0340.1EA
0
EUR
ROTH0340.1
ROTH0340.2
ROTH0340.3
(±)-1,2-Propanediol
Propylene glycol
Formula:
CH₃CH(OH)CH₂OH MW: 76.1 g/mol Boiling Pt: 187.6 Melting Pt: –59 °C Density: 1.04 g/cm³ (20 °C) Flash Pt: 99 Storage Temperature: Ambient |
MDL Number:
MFCD00064272 CAS Number: 57-55-6 EINECS: 200-338-0 Merck Index: 13,07947 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...