(±)-1-Indanol 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(±)-1-Hydroxyindan, 1-Indanol, 2,3-Dihydro-1H-inden-1-ol
186110100
186110500
ACRO186110100EA
112
EUR
ACRO186110100
ACRO186110500
(±)-1-Indanol 98%
(±)-1-Indanol
Formula:
C₉H₁₀O MW: 134,18 g/mol Melting Pt: 50…54 °C Flash Pt: 145 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003797 CAS Number: 6351-10-6 EINECS: 228-755-3 UN: 2811 |
Specification Test Results
Appearance | White to yellow to beige to light brown Powder or crystals and/or chunks or fused solid |
Infrared spectrum | Conforms |
Melting point | 50 to 55 °C |
GC | ≥97.5 % |
HPLC | ≥97.5 % |
Specific optical rotation | +7° to +10° (20 °C, 589 nm) (1% in MeOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...