Sodium carbonate decahydrate

Supplier: Roth Carl
Warning

8566.1 8566.2 8566.3
ROTH8566.1EA 0 EUR
ROTH8566.1 ROTH8566.2 ROTH8566.3
Sodium carbonate decahydrate
Sodium carbonate decahydrate
Formula: Na₂CO₃·10H₂O
Melting Pt: 33 °C
Density: 1,46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR