Tetrasodium diphosphate decahydrate
Supplier: Roth Carl
|
Warning
|
Synonyms:
Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
T883.1
T883.2
ROTHT883.1EA
83.9
EUR
ROTHT883.1
ROTHT883.2
Tetrasodium diphosphate decahydrate
tetra-Sodium diphosphate decahydrate
Formula:
Na₄P₂O₇·10H₂O MW: 446,06 g/mol Melting Pt: 990 °C Density: 1,82 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149200 CAS Number: 13472-36-1 EINECS: 231-767-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...