Sodium thiosulphate pentahydrate
Supplier: Roth Carl
T109.1
T109.2
ROTHT109.1EA
79.4
EUR
ROTHT109.1
ROTHT109.2
Sodium thiosulphate pentahydrate
Sodium thiosulphate pentahydrate
Formula:
Na₂S₂O₃.5H₂O MW: 248.19 g/mol Melting Pt: 48 Density: 1.67 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149186 CAS Number: 10102-17-7 EINECS: 231-867-5 Merck Index: 12,08844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...