Barium chloride dihydrate

Supplier: Molekula
Danger

Synonyms: Barium dichloride

MOLE12034507EA 0 EUR
MOLE12034507
Barium chloride dihydrate
Barium chloride dihydrate
Formula: BaCl₂·2H₂O
MW: 244,27 g/mol
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 960 °C
Density: 3,86 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 13,00974

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR