Calcium chloride dihydrate
Supplier: Roth Carl
|
Warning
|
5239.1
5239.2
ROTH5239.1EA
0
EUR
ROTH5239.1
ROTH5239.2
Calcium chloride dihydrate
Calcium chloride dihydrate
Formula:
CaCl₂·2H₂O Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 176 °C Density: 1,85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149613 CAS Number: 10035-04-8 EINECS: 233-140-8 Merck Index: 13,01660 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...