Iodocyclopentane ≥97% stabilised

Supplier: Thermo Fisher Scientific

L01522.06 L01522.14
L01522.06EA 42.5 EUR
L01522.06 L01522.14
Iodocyclopentane ≥97% stabilised
Iodocyclopentane

Stabilized with copper


Purity: 97% CAS No.: 1556-18-9 Molecular Formula: C5H9I Formula Weight: 196.03 Flash Point: 59°(138°F) Refractive Index: 1.5480 Boiling Point: 166-167° Beilstein Reference: 1900558 UN No.: 1993 MDL No.: MFCD00039446

Formula: C₅H₉I
MDL Number: MFCD00039446
CAS Number: 1556-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR