1,5-Dibromopentane ≥98%

Supplier: Thermo Fisher Scientific
Warning

A18465.0E A18465.22 A18465.36
A18465.0EEA 472 EUR
A18465.0E A18465.22 A18465.36
1,5-Dibromopentane ≥98%
1,5-Dibromopentane

Purity: 98% CAS No.: 111-24-0 Molecular Formula: Br(CH2)5Br Formula Weight: 229.95 Flash Point: 79°(174°F) Refractive Index: 1.5122 Melting Point: -34° Boiling Point: 224° Beilstein Reference: 1209245 MDL No.: MFCD00000268

Formula: BrH₂C(CH₂)₃CH₂Br
MW: 229.94 g/mol
Boiling Pt: 224 °C
Melting Pt: –39 °C
Density: 1.688
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000268
CAS Number: 111-24-0
EINECS: 203-849-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR