1-Bromooctane 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
166815000
166810025
ACRO166815000EA
109
EUR
ACRO166815000
ACRO166810025
1-Bromooctane 99%
1-Bromooctane
Formula:
BrH₂C(CH₂)₆CH₃ Boiling Pt: 201 °C (760 mmHg) Melting Pt: –55 °C Density: 1,11 g/cm³ Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000276 CAS Number: 111-83-1 EINECS: 203-912-9 UN: 3082 ADR: 9,III Merck Index: 12,06861 |
Specification Test Results
Appearance | Clear colorless to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4508 to 1.4528 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...