1-Bromoundecane ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14333.14 A14333.22 A14333.36
A14333.14EA 40.6 EUR
A14333.14 A14333.22 A14333.36
1-Bromoundecane ≥98%
1-Bromoundecane

Purity: 98% CAS No.: 693-67-4 Molecular Formula: CH3(CH2)10Br Formula Weight: 235.22 Flash Point: 110°(230°F) Refractive Index: 1.4563 Melting Point: -9° Boiling Point: 112-114°/10mm Beilstein Reference: 1098390 MDL No.: MFCD00000223

Formula: BrH₂C(CH₂)₉CH₃
Boiling Pt: 112…114 °C (10 mmHg)
Melting Pt: –9 °C
Density: 1,052
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000223
CAS Number: 693-67-4
EINECS: 211-757-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR